MAT048696457 Matrix Scientific

N-Methyl-N-methoxydifluoroacetamide

  • MDL number MFCD06248152
  • Cas Number : 142492-01-1
  • Molecular Weight : 139.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Boiling point 40�7mm
Product Link http://www.matrixscientific.com/012611.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/012611.html
Availability details for
Quantity
  • Availability
  • Add to Cart