MAT048696769 Matrix Scientific

3-{[(4-Fluorophenethyl)oxy]methyl}piperidine

  • MDL number MFCD06247629
  • Cas Number : 883537-91-5
  • Molecular Weight : 237.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/014539.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/014539.html
Availability details for
Quantity
  • Availability
  • Add to Cart