MAT048696813 Matrix Scientific

1-(4-Methoxyphenyl)-2-(pyrimidin-4-yl)ethanol

  • MDL number MFCD03426057
  • Cas Number : 185848-10-6
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-111
Country of Origin USA
Product Link http://www.matrixscientific.com/007301.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007301.html
Availability details for
Quantity
  • Availability
  • Add to Cart