MAT048697417 Matrix Scientific

3-[(Neopentyloxy)methyl]piperidine

  • MDL number MFCD06248580
  • Cas Number : 883544-53-4
  • Molecular Weight : 185.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/014914.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/014914.html
Availability details for
Quantity
  • Availability
  • Add to Cart