MAT048697488 Matrix Scientific

(4-Perfluorotolyl)triethoxysilane

  • MDL number MFCD06248661
  • Cas Number : 61069-04-3
  • Molecular Weight : 380.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.29
Country of Origin USA
Boiling point 68-70�5mm
Product Link http://www.matrixscientific.com/012889.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012889.html
Availability details for
Quantity
  • Availability
  • Add to Cart