MAT048697629 Matrix Scientific

4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone

  • MDL number MFCD00143378
  • Cas Number : 1691-93-6
  • Molecular Weight : 270.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144
Country of Origin USA
Product Link http://www.matrixscientific.com/005935.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart