MAT048697696 Matrix Scientific

5,5,6,6,6-Pentafluorohexane-2,4-dione

  • MDL number MFCD00466808
  • Cas Number : 356-40-1
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 111-112�1 Torr
Product Link http://www.matrixscientific.com/012863.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012863.html
Availability details for
Quantity
  • Availability
  • Add to Cart