MAT048697699 Matrix Scientific

3-(Pentafluorophenyl)propionic acid

  • MDL number MFCD00020516
  • Cas Number : 2002-92-8
  • Molecular Weight : 240.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Country of Origin USA
Product Link http://www.matrixscientific.com/004421.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/004421.html
Availability details for
Quantity
  • Availability
  • Add to Cart