MAT048697719 Matrix Scientific

(2-Oxo-4-phenyl-pyrrolidin-1-yl)-acetic acid

  • MDL number MFCD00453675
  • Cas Number : 67118-34-7
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-163
Country of Origin USA
Product Link http://www.matrixscientific.com/012770.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012770.html
Availability details for
Quantity
  • Availability
  • Add to Cart