MAT048697737 Matrix Scientific

3-Oxo-2-pyridin-4-ylmethyl-2,3-dihydro-1H-isoindole-4-carboxylic acid

  • MDL number MFCD06011741
  • Cas Number : 842974-44-1
  • Molecular Weight : 268.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012814.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012814.html
Availability details for
Quantity
  • Availability
  • Add to Cart