MAT048697749 Matrix Scientific

3,4,5-Trifluoroiodobenzene

  • MDL number MFCD03265259
  • Cas Number : 170112-66-0
  • Molecular Weight : 257.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerate/dark
Country of Origin USA
Product Link http://www.matrixscientific.com/012627.html

Safety Information

hcodes Irritant/Light Sensitive/Refrigerate
MSDS http://www.matrixscientific.com/012627.html
Availability details for
Quantity
  • Availability
  • Add to Cart