MAT048697761 Matrix Scientific

1-Phenyl-2-(pyrimidin-4-yl)ethanol

  • MDL number MFCD03426053
  • Cas Number : 36914-71-3
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-117
Country of Origin USA
Product Link http://www.matrixscientific.com/007291.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007291.html
Availability details for
Quantity
  • Availability
  • Add to Cart