MAT048697782 Matrix Scientific

2-(4-Methylsulfonyl-phenyl)-ethylaminehydrochloride

  • MDL number MFCD06245499
  • Cas Number : 849020-96-8
  • Molecular Weight : 235.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-205
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/012554.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012554.html
Availability details for
Quantity
  • Availability
  • Add to Cart