MAT048697802 Matrix Scientific

4,4,4-Trifluoro-3-(trifluoromethyl)crotonic acid

  • MDL number MFCD00013574
  • Cas Number : 1763-28-6
  • Molecular Weight : 208.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 90� Torr
Product Link http://www.matrixscientific.com/003581.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/003581.html
Availability details for
Quantity
  • Availability
  • Add to Cart