MAT062351249 Matrix Scientific

[2-Amino-4-(4-ethyl-phenyl)-thiazol-5-yl]-acetic acid hydrochloride

  • MDL number MFCD06799737
  • Cas Number : 1185292-96-9
  • Molecular Weight : 298.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010806.html

Safety Information

MSDS http://www.matrixscientific.com/010806.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart