MAT062351263 Matrix Scientific

2-Aminothiazole-4-carboxylic acid, ethyl esterhydrochloride

  • MDL number MFCD00859544
  • Cas Number : 435342-17-9
  • Molecular Weight : 208.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007879.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007879.html
Availability details for
Quantity
  • Availability
  • Add to Cart