MAT062351296 Matrix Scientific

Benzo[1,3]dioxol-5-ylmethyl-pyridin-4-ylmethyl-amine hydrobromide

  • MDL number MFCD06799784
  • Cas Number : 1185298-32-1
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011590.html

Safety Information

MSDS http://www.matrixscientific.com/011590.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart