MAT062351469 Matrix Scientific

4-(4-Benzyl-1-piperazino)-2-bromo-benzaldehyde

  • MDL number MFCD06656552
  • Cas Number : 883512-11-6
  • Molecular Weight : 359.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016620.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016620.html
Availability details for
Quantity
  • Availability
  • Add to Cart