MAT062351486 Matrix Scientific

1,4'-Bipiperidine-3-carboxylic aciddihydrochloride

  • MDL number MFCD06799961
  • Cas Number : 1185293-24-6
  • Molecular Weight : 285.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016329.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016329.html
Availability details for
Quantity
  • Availability
  • Add to Cart