MAT062351489 Matrix Scientific

(1,4'-Bipiperidin-4-ylmethyl)dimethylamine

  • MDL number MFCD06799964
  • Cas Number : 883516-04-9
  • Molecular Weight : 225.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016371.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016371.html
Availability details for
Quantity
  • Availability
  • Add to Cart