MAT062351500 Matrix Scientific

3,5-Bis(trifluoromethyl)phenylhydrazinehydrochloride

  • MDL number MFCD03094165
  • Cas Number : 502496-23-3
  • Molecular Weight : 280.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/005834.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005834.html
Availability details for
Quantity
  • Availability
  • Add to Cart