MAT062351722 Matrix Scientific

3-Bromo-N-(2-fluorophenyl)-4-(tetrahydro-2-furanylmethoxy)benzamide

  • MDL number MFCD06800185
  • Cas Number : 883515-08-0
  • Molecular Weight : 394.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/017892.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/017892.html
Availability details for
Quantity
  • Availability
  • Add to Cart