MAT062351848 Matrix Scientific

4-(Allyloxy)aniline

  • MDL number MFCD01902260
  • Cas Number : 1688-69-3
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 142-143� Torr
Product Link http://www.matrixscientific.com/017129.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/017129.html
Availability details for
Quantity
  • Availability
  • Add to Cart