MAT062351850 Matrix Scientific

1-(Aminocarbonyl)piperidine-4-carboxylic acid

  • MDL number MFCD06655891
  • Cas Number : 851168-77-9
  • Molecular Weight : 172.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016333.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016333.html
Availability details for
Quantity
  • Availability
  • Add to Cart