MAT062351864 Matrix Scientific

1-(2-Bromoethoxy)-4-fluorobenzene

  • MDL number MFCD00044739
  • Cas Number : 332-48-9
  • Molecular Weight : 219.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-60
Country of Origin USA
Boiling point 123� Torr
Product Link http://www.matrixscientific.com/016381.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016381.html
Availability details for
Quantity
  • Availability
  • Add to Cart