MAT062351882 Matrix Scientific

3-Amino-N,N-dimethyl-benzenesulfonamide

  • MDL number MFCD00602451
  • Cas Number : 6274-18-6
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016779.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016779.html
Availability details for
Quantity
  • Availability
  • Add to Cart