MAT062351888 Matrix Scientific

1-Amino-3,3-dimethyl-butan-2-onehydrobromide

  • MDL number MFCD07277376
  • Cas Number : 77428-85-4
  • Molecular Weight : 196.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010263.html

Safety Information

MSDS http://www.matrixscientific.com/010263.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart