MAT062351927 Matrix Scientific

4-(1-Adamantyl)-2-aminothiazolehydrochloride

  • MDL number MFCD01825559
  • Cas Number : 462605-77-2
  • Molecular Weight : 270.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011487.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/011487.html
Availability details for
Quantity
  • Availability
  • Add to Cart