MAT062352025 Matrix Scientific

Allyl-but-2-ynyl-amine hydrochloride

  • MDL number MFCD03596897
  • Cas Number : 436099-58-0
  • Molecular Weight : 145.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008660.html

Safety Information

MSDS http://www.matrixscientific.com/008660.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart