MAT062352045 Matrix Scientific

3-Amino-3-pyridin-3-ylpropanoic acid

  • MDL number MFCD01549598
  • Cas Number : 62247-21-6
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206
Country of Origin USA
Product Link http://www.matrixscientific.com/016494.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016494.html
Availability details for
Quantity
  • Availability
  • Add to Cart