MAT062352046 Matrix Scientific

3-Amino-3-benzo[1,3]dioxol-5-yl-propionic acidethyl ester hydrochloride

  • MDL number MFCD05982931
  • Cas Number : 149498-94-2
  • Molecular Weight : 273.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-183
Country of Origin USA
Product Link http://www.matrixscientific.com/011931.html

Safety Information

MSDS http://www.matrixscientific.com/011931.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart