MAT062352085 Matrix Scientific

4-(2-Amino-1,3-thiazol-4-yl)benzene-1,3-diolhydrochloride

  • MDL number MFCD01849140
  • Cas Number : 472979-20-7
  • Molecular Weight : 244.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011645.html

Safety Information

MSDS http://www.matrixscientific.com/011645.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart