MAT062352086 Matrix Scientific

4-(2-Amino-1,3-thiazol-4-yl)-2-methoxyphenolhydrochloride

  • MDL number MFCD01897969
  • Cas Number : 1056451-14-9
  • Molecular Weight : 258.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011730.html

Safety Information

MSDS http://www.matrixscientific.com/011730.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart