MAT062352886 Matrix Scientific

5-Chloro-1-methyl-1H-indole-2-carbaldehyde

  • MDL number MFCD06800429
  • Cas Number : 883529-71-3
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016217.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016217.html
Availability details for
Quantity
  • Availability
  • Add to Cart