MAT062352888 Matrix Scientific

[(6-Chloro-7-methyl-1H-indol-2-yl)-methyl]amine

  • MDL number MFCD06800431
  • Cas Number : 883529-85-9
  • Molecular Weight : 194.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016181.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016181.html
Availability details for
Quantity
  • Availability
  • Add to Cart