MAT062352891 Matrix Scientific

5-Chloromethyl-2,2,8-trimethyl-4H-[1,3]dioxino-[4,5-c]pyridine hydrochloride

  • MDL number MFCD00449590
  • Cas Number : 6562-92-1
  • Molecular Weight : 264.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-187
Country of Origin USA
Product Link http://www.matrixscientific.com/007904.html

Safety Information

MSDS http://www.matrixscientific.com/007904.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart