MAT062352901 Matrix Scientific

7-Chloro-4-(trifluoromethyl)-1H-indole-2-carbaldehyde

  • MDL number MFCD06800444
  • Cas Number : 883522-93-8
  • Molecular Weight : 247.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016203.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016203.html
Availability details for
Quantity
  • Availability
  • Add to Cart