MAT062352908 Matrix Scientific

1-(4-Cyanobenzyl)-1H-indole-2-carboxylic acid

  • MDL number MFCD06800449
  • Cas Number : 229953-48-4
  • Molecular Weight : 276.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016231.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016231.html
Availability details for
Quantity
  • Availability
  • Add to Cart