MAT062352940 Matrix Scientific

[(1-Cyclohexylpiperidin-3-yl)methyl]amine

  • MDL number MFCD06800482
  • Cas Number : 883533-78-6
  • Molecular Weight : 196.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016361.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016361.html
Availability details for
Quantity
  • Availability
  • Add to Cart