MAT062352944 Matrix Scientific

Cyclohexyl-pyridin-3-ylmethyl-aminehydrochloride

  • MDL number MFCD06800485
  • Cas Number : 1135239-09-6
  • Molecular Weight : 226.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010235.html

Safety Information

MSDS http://www.matrixscientific.com/010235.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart