MAT062353019 Matrix Scientific

4,4-Difluoro-3-butenylacetate

  • MDL number MFCD06800562
  • Cas Number : 929-12-4
  • Molecular Weight : 150.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 123
Product Link http://www.matrixscientific.com/018208.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018208.html
Availability details for
Quantity
  • Availability
  • Add to Cart