MAT062353029 Matrix Scientific

C-(2,3-Dihydro-benzofuran-2-yl)-methylaminehydrochloride

  • MDL number MFCD01659957
  • Cas Number : 19997-54-7
  • Molecular Weight : 185.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010859.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010859.html
Availability details for
Quantity
  • Availability
  • Add to Cart