MAT062353038 Matrix Scientific

3-(3,4-Dihydro-2H-quinolin-1-yl)-propionic acidhydrochloride

  • MDL number MFCD06800578
  • Cas Number : 884048-30-0
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171
Country of Origin USA
Product Link http://www.matrixscientific.com/007864.html

Safety Information

MSDS http://www.matrixscientific.com/007864.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart