MAT062353052 Matrix Scientific

(2,4-Dimethoxy-benzyl)-pyridin-4-ylmethyl-aminehydrochloride

  • MDL number MFCD06800587
  • Cas Number : 1135233-39-4
  • Molecular Weight : 294.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011596.html

Safety Information

MSDS http://www.matrixscientific.com/011596.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart