MAT062353053 Matrix Scientific

(3,4-Dimethoxy-benzyl)-pyridin-4-ylmethyl-aminehydrochloride

  • MDL number MFCD06800588
  • Cas Number : 1185294-28-3
  • Molecular Weight : 294.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011597.html

Safety Information

MSDS http://www.matrixscientific.com/011597.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart