MAT062353061 Matrix Scientific

2,7-Dimethoxy-3,6-bis(methylthio)-naphthalene

  • MDL number MFCD06656527
  • Cas Number : 105404-97-5
  • Molecular Weight : 280.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016593.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016593.html
Availability details for
Quantity
  • Availability
  • Add to Cart