MAT062353069 Matrix Scientific

3,4-Dimethoxyphenylglyoxal hydrate

  • MDL number MFCD06800598
  • Cas Number : 1138011-18-3
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-91
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/016074.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016074.html
Availability details for
Quantity
  • Availability
  • Add to Cart