MAT062353078 Matrix Scientific

2',4'-Dimethyl[4,5']bithiazolyl-2-ylaminehydrobromide

  • MDL number MFCD01419978
  • Cas Number : 435341-84-7
  • Molecular Weight : 292.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007950.html

Safety Information

MSDS http://www.matrixscientific.com/007950.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart