MAT062353094 Matrix Scientific

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde

  • MDL number MFCD06800625
  • Cas Number : 934570-54-4
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016275.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016275.html
Availability details for
Quantity
  • Availability
  • Add to Cart