MAT062353099 Matrix Scientific

[3-(3,5-Dimethyl-1H-pyrazol-1-yl)-benzyl]methylamine dihydrochloride

  • MDL number MFCD06800630
  • Cas Number : 1185293-07-5
  • Molecular Weight : 288.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016278.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016278.html
Availability details for
Quantity
  • Availability
  • Add to Cart